Applikon Reagent Addition Bottles
Reagent Addition Bottles come in two sizes. Use small bottle for acids or bases; use large bottle to hold fresh media. Transfer pipes and filter included.
Autoclavable.
$682.00 - $1,280.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Applikon Fermentation Reagent Addition Bottle, 1.0 L EW-01919-91 Mfr # Z811301910
COMPARE
Fermentation Reagent Addition Bottle, 1.0 L
$682.00USD / Each
Applikon Fermentation Reagent Addition Bottle, 5.0 L EW-01919-92 Mfr # Z811301912
COMPARE
Fermentation Reagent Addition Bottle, 5.0 L
$1,280.00USD / Each