Analog Balancing Balometer
The only analog capture hood in production on the planet
  • Three measuring ranges: 0 -250, 200 – 500, and 400 – 1000 CFM
The Alnor® ABT® Balometer is a high productivity instrument designed to quickly measure the airflow directly from supply diffusers and return/exhaust grilles. The ABT consists of a fabric hood, hood frame, support rods, and a molded plastic base which contains the analog meter and flow sensing manifold.
The ABT models are temperature-compensated to display a standard flow rate in CFM. Standard flow rate is defined as the volumetric flow rate at standard conditions of 70°F (21.1°C) and 14.7 psi (760 mmHg) barometric pressure.
For increased flexibility, additional hood sizes are available to fit interchangeably on the instrument base.
$3,294.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Max Air Volume (CFM) Volume Accuracy Volume Resolution
Availability Pricing
TSI ABT701 Analog Balometer Kit, CFM with 2’ x 2’ hood EW-68511-50 Mfr # ABT701
COMPARE
1000 ±3% full scale + 5 CFM 5, 10, 20, 400 CFM
$3,294.00USD / Each