Amprobe Relative Humidity Temperature Meters
The meters use a precision capacitance sensor to deliver accurate performance with long term stability.
  • Dual display
  • Relative function monitors humidity and temperature changes
  • Extended probe stem to easily access air ducts
  • Min / Max readings
  • Data Hold
  • Capacitive sensor with protective cover
  • Auto Power Off
  • Wet Bulb / Dewpoint measurements (THWD-3 only)
This high accuracy instrument measures the full range of relative humidity from 0% to 100% and features an exceptionally wide ambient temperature measurement range from –4degF to 140degF ( -20degC to
60degC). The sensor is mounted on a long shaft extending from the top surface of the unit, making probing into ductwork and hard to reach areas easier. The large LCD displays two measurements simultaneously with a full range of display options including Hold, Min, Max and Relative. The THWD-3 also measures dewpoint and wet bulb.
ITEM(S) ON SALE
$212.39 - $224.09USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Battery Max RH Range (%) Max Temperature (° F)
Availability Pricing
Amprobe TH-3 Relative Humidity Temperature Meter, 2 Display EW-10508-36 Mfr # TH-3
COMPARE
1 9V (included) 100 140
$212.39USD / Each
$235.99USD / Each
Amprobe THWD-3 Relative Humidity Temperature Meter, Dew Point, Wet Bulb EW-10508-37 Mfr # THWD-3
COMPARE
1 9V (included) 100 140
$224.09USD / Each
$248.99USD / Each