Amprobe Digital Light Meters, Lux/Foot-candles
Reduce energy costs by increasing lighting system efficiencies
  • Data hold function freezes displayed measurements
  • Min/max capability for high and low measurements
  • Automatic, positive implied, and negative polarity indication
  • Auto power-off saves battery life
  • Cosine angular corrected per JIS C 1609:1993 and CNS 5119 general A class
  • Auto-ranging plus manual range (model LM-120)
  • Zero function nulls display before measurement (model LM-120)
These digital light meters measure visible light from fluorescent, metal halide, high-pressure sodium, or incandescent sources. Meters include a silicon photodiode sensor and filter, and feature a wide measuring range with precision accuracy (0.01 Fc/Lux) making them ideal for measuring output levels of lighting fixtures and more. Meters are designed for simple one-hand operation and feature a large easy-to-read 3-1/2 digital LCD display.
ITEM(S) ON SALE
$194.39 - $243.89USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Function
Availability Pricing
Amprobe LM-100 Digital Light Meter; 0 to 20,000 Footcandles EW-50512-50 Mfr # LM-100
COMPARE
Data hold, min/max, manual ranging, auto power-off, positive implied and negative polarity indication
$194.39USD / Each
$215.99USD / Each
Amprobe LM-120 Autoranging Digital Light Meter EW-50512-51 Mfr # LM-120
COMPARE
Data hold, min/max, autoranging, autozero, auto power-off, positive implied and negative polarity indication
$243.89USD / Each
$270.99USD / Each