Allsafe/SMC MonoGoggle XTR Goggles
Greater range of visibility with a more comfortable fit
Lightweight goggle provides superior impact and chemical splash protection. Cylindrical-shaped lens offers greater range of visibility. The 0.060"-thick polycarbonate lens features antifog coating and provides 99.9% UV protection. The soft frame is pliable and conforms to worker’s face for a comfortable fit.
Compliance: ANSI Z87.1+ (High impact)
$15.40 - $36.10USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Frame Color Lens Color Description
Availability Pricing
Allsafe 3010334 Blue Frame, Clear Lens EW-81657-15 Mfr # 18624
COMPARE
Blue Clear Blue Frame, Clear Lens
$15.40USD / Each
Allsafe 3010343 Shield, Blue Frame, Clear Lens EW-81657-18 Mfr # 18629
COMPARE
Blue Clear Shield, Blue Frame, Clear Lens
$36.10USD / Each