Airgas® Radnor® Disposable Earplugs
Hearing protection for the loudest work environments
  • Soft polyurethane foam construction
  • Soil-resistant skin
  • NBR 29 dB rated
  • Intense orange plugs and optional cord are highly visible
Airgas® Radnor® disposable earplugs are constructed of polyurethane foam that is light, soft, and comfortable in your ear canal. It has a smooth, soil-resistant skin to prevent dirt build-up. The easy to roll-down design resists the tendency to back out of the ear canal, ensuring a tight seal for your protection. Disposable earplugs are NBR 29 dB compliant and are available corded or uncorded.
$21.60 - $22.80USD / Box of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Corded NRR (dB) Description
Availability Pricing
Radnor Tapered Polyurethane Foam Disposable Uncorded Earplugs; 200 Pairs/Box
New Products
EW-78900-65 Mfr # RAD64051810
COMPARE
No 29 Tapered Polyurethane Foam Disposable Uncorded Earplugs; 200 Pairs/Box
$21.60USD / Box of 200
Radnor Tapered Polyurethane Foam Disposable Corded Earplugs; 100 Pairs/Box
New Products
EW-78900-66 Mfr # RAD64051811
COMPARE
Yes 29 Tapered Polyurethane Foam Disposable Corded Earplugs; 100 Pairs/Box
$22.80USD / Box of 100