This product is now discontinued, View Related Products
Cole-Parmer® Essentials Vacuum/Pressure Pumps Accessories
Dual-head pump gives you increased capacity over the single-head AIR CADET pumps. Use the two pump heads independently or connect them in series with 3/8" ID Tygonr vacuum tubing for up to 23" Hg vacuum.
Dual-head pump includes thread tape made of PTFE PTFE resin, four polyethylene 3/8" NPT(M) x 3/8" hose barb adapters, and a 6-ft, three-wire cord; 115 VAC models include a plug.
Call For Price
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Brass vacuum/pressure gauge connector kit for all pumps