AEMC 4610, 4620, 4630 Digital Ground Resistance Testers
Easy-to-operate: connect, press and read
  • Measures soil resistivity (4-Point) method
  • Measures ground resistance (2- and 3-Point) Fall-of-Potential method
  • Step voltage tests and touch potential measurements
  • Auto-Ranging: automatically selects the optimum range
  • Designed to reject high levels of noise interference
LED on faceplate informs operator of high input noise, high auxiliary rod resistance and fault connections. Rugged, dustproof and rainproof field case. Can also be used for continuity tests on bonding.
1 PRODUCT OPTIONS
Compareitem specifications
AC Current Accuracy AC Current Range (mA) AC Current Resolution (mA)
Availability Pricing
AEMC 4620KIT-500FT Kit-500 ft Ground Resistance Tester EW-20015-05 Mfr # 2135.21
COMPARE
10mA, 1mA, 0.1mA
$2,041.55USD / Each