Advantec Ion Test Strips
Color-change test papers give rapid indication of pH level ion presence
  • Obtain quick ion concentration measurements in the field or laboratory
  • Dip and compare color change — no calibration, electricity, or warm up needed
These ion test strips make water testing fast and easy. Conveniently packed in screw top bottles, the test papers are safe even in rugged field applications. Obtaining simultanious pH and ion concentration results couldn't be easier. Simply dip the test strip in your water sample, wait about 15 seconds for the water to penetrate the strip, and compare the color changes against the chart on the bottle's label.
$19.20 - $90.90USD / Pkg of 25
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Measurement Points Test Tests Per Kit
Availability Pricing
Advantec 07030010 Test Strips, Alumi Check (Aluminum Al3+), 0 to 100 ppm; 100 Strips/Pk EW-59201-10 Mfr # 07030010
COMPARE
0 to 100 ppm Aluminum 100
$90.90USD / Pkg of 100
Advantec 07030020 Test Strips, Copper Check (Cu2+), 0 to 500 ppm; 100 Strips/Pk EW-59201-12 Mfr # 07030020
COMPARE
0 to 500 ppm Copper, Cu2+ 100
$64.60USD / Pkg of 100
Advantec 07030030 Test Strips, Copper Check A (Cu+ and Cu2+), 0 to 50 ppm; 100 Strips/Pk EW-59201-14 Mfr # 07030030
COMPARE
0 to 50 ppm Copper, Cu+ and Cu2+ 100
$60.90USD / Pkg of 100
Advantec 07030041 Test Strips, Chrome Check (Cr6+), 0 to 200 ppm; 25 Strips/Pk EW-59201-16 Mfr # 07030041
COMPARE
0 to 200 ppm Chrome 25
$22.40USD / Pkg of 25
Advantec 07030051 Test Strips, Chrome Check A (Cr6+), 0 to 50 ppm; 25 Strips/Pk EW-59201-18 Mfr # 07030051
COMPARE
0 to 50 ppm Chrome 25
$19.20USD / Pkg of 25
Advantec 07030060 Test StripsIron Check (Fe2+), 0 to 1000 ppm; 100 Strips/Pk EW-59201-20 Mfr # 07030060
COMPARE
0 to 1000 ppm Iron 100
$62.00USD / Pkg of 100
Advantec 07030070 Test Strips, Nickel Check (Ni2+), 0 to 1000 ppm; 100 Strips/Pk
Always in Stock
EW-59201-22 Mfr # 07030070
COMPARE
0 to 1000 ppm Nickel 100
$62.00USD / Pkg of 100
Advantec 07030100 Test Strips, Cyanogen Check (Cyanide Cn-), 0 to 1000 ppm; 100 Strips/Pk
Always in Stock
EW-59201-24 Mfr # 07030100
COMPARE
0 to 1000 ppm Cyanogen 100
$62.00USD / Pkg of 100
Showing 1- 8 of 8