Additel Hydraulic Test Pumps
Unique screw press technology for easy operation and no leakage
  • Screw press technology eliminates need for built-in nonreturning valve
  • Unique quick-connect fittings tighten easily by hand, no tools or tape needed
  • High pressure generated easily and adjusted smoothly
  • Five models to meet your application and budget
$2,018.00 - $5,041.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Pressure Description
Availability Pricing
Additel 925-N Handheld Hydraulic Test Pump, 85% Vacuum to 6000 psi EW-16106-06 Mfr # ADT925-N
COMPARE
6000 psi (400 bar) Handheld Hydraulic Test Pump, 85% Vacuum to 6000 psi
$2,018.00USD / Each
Additel 927-N Portable Hydraulic Test Pump, 85% Vacuum to 10,000 psi EW-16106-07 Mfr # ADT927-N
COMPARE
10000 psi (700 bar) Portable Hydraulic Test Pump, 85% Vacuum to 10,000 psi
$2,652.00USD / Each
Additel 936-N Bench-top Hydraulic Calibration Pump, 85% Vacuum to 15,000 psi EW-16106-08 Mfr # ADT936-N
COMPARE
15000 psi (1000 bar) Bench-top Hydraulic Calibration Pump, 85% Vacuum to 15,000 psi
$5,041.00USD / Each