Additel High Accuracy Digital Pressure Gauges with Silicon Pressure Sensors
Get 0.1% Reading Accuracy
  • % pressure indication for quick visual reference
  • Flash warning if pressure goes over 120% full-scale
  • Eleven selectable pressure units
  • Large easy-to-read display with 5-digit resolution
  • Backlight for easy viewing in dimly lit areas
Now available with 0.1% of reading accuracy, the 681 series digital pressure gauges offer an economic solution for a wide variety of pressure measurement applications. Designed with state-of-the-art silicon pressure sensors, the gauges feature fully temperature compensated accuracy, programmable pressure alarm, over pressure warning, and minimum/maximum pressure display. Real-time data logging and graphical pressure readings can be recorded and dowloaded through the RS-232 port using Additel/Log II software (68336-91).
$1,304.00 - $1,413.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Additel 681 Digital Pressure Gauge, 0 to 10 psi, .25" NPT EW-68336-01 Mfr # ADT681-RD-GP10-PSI-N
COMPARE
Digital Pressure Gauge, 0 to 10 psi, .25\" NPT
$1,304.00USD / Each
Additel 681 Digital Pressure Gauge, 0 to 15 psi, .25" NPT EW-68336-03 Mfr # ADT681-RD-GP15-PSI-N
COMPARE
Digital Pressure Gauge, 0 to 15 psi, .25\" NPT
$1,304.00USD / Each
Additel 681 Digital Pressure Gauge, 0 to 50 psi, .25" NPT EW-68336-07 Mfr # ADT681-RD-GP50-PSI-N
COMPARE
Digital Pressure Gauge, 0 to 50 psi, .25\" NPT
$1,304.00USD / Each
Additel 681 Digital Pressure Gauge, 0 to 300 psi, .25" NPT EW-68336-11 Mfr # ADT681-RD-GP300-PSI-N
COMPARE
Digital Pressure Gauge, 0 to 300 psi, .25\" NPT
$1,304.00USD / Each
Additel 681 Digital Pressure Gauge, 0 to 600 psi, .25" NPT EW-68336-15 Mfr # ADT681-RD-GP600-PSI-N
COMPARE
Digital Pressure Gauge, 0 to 600 psi, .25\" NPT
$1,304.00USD / Each
Additel 681 Digital Pressure Gauge, 0 to 1 K psi, .25" NPT EW-68336-17 Mfr # ADT681-RD-GP1K-PSI-N
COMPARE
Digital Pressure Gauge, 0 to 1 K psi, .25\" NPT
$1,304.00USD / Each
Additel 681 Digital Pressure Gauge, 0 to 2 K psi, .25" NPT EW-68336-19 Mfr # ADT681-RD-GP2K-PSI-N
COMPARE
Digital Pressure Gauge, 0 to 2 K psi, .25\" NPT
$1,304.00USD / Each
Additel 681 Digital Pressure Gauge, 0 to 10 K psi, .25" NPT EW-68336-25 Mfr # ADT681-RD-GP10K-PSI-N
COMPARE
Digital Pressure Gauge, 0 to 10 K psi, .25\" NPT
$1,413.00USD / Each
Showing 1- 8 of 8