Cole-Parmer® Spex® Adapters and Clamps for High-Energy Mixer/Mill® Grinders
Use adapters to run multiple samples or to hold vial securely in place during grinding
  • Suitable for use with the Cole-Parmer® Freezer/Mill® Cryogenic Grinders
Optimize performance of your Cole-Parmer® Freezer/Mill® grinder with a wide range of adapters and clamps.
$105.00 - $197.50USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Compatible With Description
Availability Pricing
COMPARE
1/2\" dia grinding vials Multiple Sample Vial Adapter, Holds Seven 1/2\" (12.7 mm) Dia Vials; 1/EA
$191.25USD / Each
COMPARE
3/4\" dia grinding vials Multiple Sample Vial Adapter, Holds Four 3/4\" (19.1 mm) Dia Vials; 1/EA
$191.25USD / Each
COMPARE
Multiple sample vial adapters04576-65 and -66 Vial Clamp Adapter for Multiple Sample Vial Adapters 04576-65 and 04576-66; 1/EA
$119.00USD / Each
COMPARE
Alumina ceramic grinding vial set 41005-86 Clamps for Slurry Grinding in the Alumina Ceramic Grinding Vial Set 41005-86; 2/ST
$197.50USD / Set of 1
COMPARE
Polypropylene grinding vial 04576-29 Adapter for 35 mL Polypropylene Grinding Vial 04576-29; 1/EA
$105.00USD / Each
COMPARE
Polypropylene grinding vial 04576-30 Adapter for 75 mL Polypropylene Grinding Vial 04576-30; 1/EA
$119.75USD / Each
COMPARE
Vial 04575-80 Multi Vial Adapter for 2 mL Vial 04575-80, Aluminum
$190.25USD / Each
COMPARE
5 mL plastic grinding vials Multi Vial Adapter, Holds 5 mL Plastic Vials, Aluminum
$190.25USD / Each
Showing 1- 8 of 8