Adam Equipment TBB Triple Beam Balances
Low cost and simple to use
  • Magnetic damping for faster stability
  • Rigid construction for durability
  • Zero adjust facility to enable zero reading
  • Notched and tiered graduated beams with centre reading
  • Stainless steel weighing pan
  • Ruler for measuring cm
  • Weight holders for storing additional weights
  • Tripod legs for density: TBB 2610S and TBB 2610T models only.
  • Large stainless steel pan
  • Lock down mounting slot for Kensington™ type lock and cable
Built tough With a larger overall body than other triple beam balances, the TBB is the sturdiest choice. Quality construction with strong metal housing and a stainless steel pan.
Ready for theft protection A factoryinstalled security slot is built right into the housing.
Better below-balance weighing Innovative tripod legs and integral weigh-below hook allow you to do density activities wherever you want without the inconvenience of having to clamp onto a table and create your own make-shift hook. (Legs are standard with 2610g models)
Reliable readings Three graduated-tier beams for the best possible centerpoint readings. Speedy zeroing with counterbalance knob.
Added value With a conversion chart, ruler, and protractor right on the housing, the TBB is a complete measurement tool
ITEM(S) ON SALE
$141.90 - $184.90USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (g) Description
Availability Pricing
Adam Equipment TBB610S Balance, 610g Capacity and 0.1g Readability EW-11710-37 Mfr # TBB 610S
COMPARE
610 0.1 Balance, 610g Capacity and 0.1g Readability
$141.90USD / Each
$165.00USD / Each
Adam Equipment TBB2610T Balance, 2610g Capacity and 0.1g Readability with Tare EW-11710-49 Mfr # TBB 2610T
COMPARE
2610 0.1 Balance, 2610g Capacity and 0.1g Readability with Tare
$184.90USD / Each
$215.00USD / Each