Adam Equipment Solis Precision Toploading Balances
Intuitive graphic display and keypad simplify weighing operations
  • Formulation and recipe function stores and recalls up to 99 ingredients
  • Selectable digital filtering minimizes effects of vibration and disturbances
  • Level indicator and adjustable feet ensure proper balance setup
  • Security slot for optional Kensington-style lock and cable prevents theft
  • Large stainless steel weighing pan allows quick cleaning
  • Time and date stamp meets Good Laboratory Practice (GLP) guidelines
The Solis line of precision toploading balances feature large high-resolution graphic displays where you can readily view your comprehensive data. Intuitive keypad, easy-to-read buttons, and navigation arrows simplify balancing operations. A built-in memory stores and recall products, formulas, and settings.
The balances also feature RS-232 interface to transfer data to computers and printers, checkweighing with audible alarm, part counting with preset sample sizes, dynamic weighing for animals or liquids, percentage weighing, density determination with optional density kit, security password protection, and multilingual display capability.
Choose from a variety of capacities with external calibration (must use calibration mass to calibrate), or internal calibration (may use with or without a calibration mass).
ITEM(S) ON SALE
$1,311.50 - $2,734.80USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Description
Availability Pricing
Adam Equipment SPB 363e Solis Precision Toploading Balance, 360g x .001g, External Calibration EW-11125-40 Mfr # SPB 363E
COMPARE
External 360 Precision Toploading Balance, 360g x .001g, External Calibration
$1,423.30USD / Each
$1,655.00USD / Each
Adam Equipment STB 3202e Solis Precision Toploading Balance, 3200g x .01g, External Calibration EW-11125-42 Mfr # STB 3202E
COMPARE
External 3200 Precision Toploading Balance, 3200g x .01g, External Calibration
$1,311.50USD / Each
$1,525.00USD / Each
Adam Equipment STB 6202e Solis Precision Toploading Balance, 6200g x .01g, External Calibration EW-11125-43 Mfr # STB 6202E
COMPARE
External 6200 Precision Toploading Balance, 6200g x .01g, External Calibration
$1,969.40USD / Each
$2,290.00USD / Each
Adam Equipment SPB 723i Solis Precision Toploading Balance, 720g x .001g, Internal Calibration EW-11125-46 Mfr # SPB 723I
COMPARE
Internal 720 Precision Toploading Balance, 720g x .001g, Internal Calibration
$1,866.20USD / Each
$2,170.00USD / Each
Adam Equipment SPB 1203i Solis Precision Toploading Balance, 1200 g x .001g, Internal Calibration EW-11125-47 Mfr # SPB 1203I
COMPARE
Internal 1200 Precision Toploading Balance, 1200 g x .001g, Internal Calibration
$2,403.70USD / Each
$2,795.00USD / Each
Adam Equipment SPB 2103i Solis Precision Toploading Balance, 2100g x .001g, Internal Calibration EW-11125-48 Mfr # SPB 2103I
COMPARE
Internal 2100 Precision Toploading Balance, 2100g x .001g, Internal Calibration
$2,734.80USD / Each
$3,180.00USD / Each
Adam Equipment STB 3202i Solis Precision Toploading Balance, 3200g x .01g, Internal Calibration EW-11125-49 Mfr # STB 3202I
COMPARE
Internal 3200 Precision Toploading Balance, 3200g x .01g, Internal Calibration
$1,526.50USD / Each
$1,775.00USD / Each
Adam Equipment STB 6202i Solis Precision Toploading Balance, 6200g x .01g, Internal Calibration EW-11125-50 Mfr # STB 6202I
COMPARE
Internal 6200 Precision Toploading Balance, 6200g x .01g, Internal Calibration
$2,188.70USD / Each
$2,545.00USD / Each
Showing 1- 8 of 8