Adam Equipment Nimbus Heavy Weight Top Loading Balances
Reliable performance in a compact, rugged design
  • All-metal housing ensures rugged, durable protection and stability
  • GLP compliant printouts via RS-232 connection
  • Weighing applications include parts counting, check weighing, percent weighing, dynamic/animal weighing
  • Below-balance weighing hook
  • Date and time stored in memory
  • Large backlit display and intuitive keypad
Optimized connections boost data collection capabilities and lead to exceptional information transmission. USB and RS-232 interfaces are standard, while an additional interface allows use of an optional remote display. Whether performing basic tasks such as printing data, or advanced communication with a LIMS system, the Nimbus can meet the requirements. GLP printouts are available with time, date and other essential information.
Innovative design creates a smaller footprint, so the Nimbus occupies minimal space on the lab bench. Solid metal construction throughout the balance durably withstands chemicals and rigorous daily use. Optimized configuration of the Nimbus’s internal weighing system results in enhanced performance. Fabricated from a single block, the weighing sensor contains fewer parts than a traditional force motor balance, improving efficiency.
ITEM(S) ON SALE
$1,797.40 - $3,160.50USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
Adam Equipment NBL 12001-115V Heavy Weight Toploader Balance, 12000g x 0.1g 115V EW-11121-24 Mfr # NBL 12001E
COMPARE
12000 115 0.1
$1,797.40USD / Each
$2,090.00USD / Each
Adam Equipment NBL 16001-115V Heavy Weight Toploader Balance, 16000g x 0.1g, 115V EW-11121-26 Mfr # NBL 16001E
COMPARE
16000 115 0.1
$2,218.80USD / Each
$2,580.00USD / Each
Adam Equipment NBL 22001-115V Heavy Weight Toploader Balance, 22000g x 0.1g 115V EW-11121-28 Mfr # NBL 22001E
COMPARE
22000 115 0.1
$3,057.30USD / Each
$3,555.00USD / Each
Adam Equipment NBL 22001-230V Heavy Weight Toploader Balance, 22000g x 0.1g, 230V EW-11121-29 Mfr # NBL 22001E
COMPARE
22000 230 0.1
$3,057.30USD / Each
$3,555.00USD / Each
Adam Equipment NBL 22001p Heavy Weight Toploader Balance, 22000 g x 0.1 g , 115 V EW-11122-56 Mfr # NBL 22001P
COMPARE
22000 115 0.1
$3,160.50USD / Each
$3,675.00USD / Each