Adam Equipment Luna Toploading Balances
Color-coded navigation buttons facilitate quick recognition
  • Level indicator and adjustable feet ensure proper balance setup
  • Sealed keypad protects against dirt and spills
  • USB and RS-232 interfaces provide fast communication with computer/printer
  • Multilingual display permits use in various regions
  • Selectable digital filtering for animal/dynamic weighing
  • Security slot for optional Kensington-type lock and cable to prevent theft
  • Auto power-off saves energy
Designed with an array of features and functions, the Luna precision toploading balances accommodate most lab applications to provide faster, more accurate readings. Functions include weighing, parts counting, percentage weighing, dynamic/animal weighing, density determination, checkweighing, and net total weighing. A backlit LCD with 24-mm digits in blue on a black background makes it is easy for you to read in any lighting conditions. Simple to operate, the navigation buttons are color-coded allowing you to perform functions with minimal training.
USB and RS-232 interfaces facilitate data collection and transmission to any computer or printer. GLP printouts are available with time, date and other essential information. Constructed of ABS plastic with a rugged 304 stainless steel pan, these balances are easy to clean and able to withstand demanding lab use. The balances offer a readability of 0.0001 g and can perform basic and detailed measuring tasks.
ITEM(S) ON SALE
$860.00 - $1,844.70USD / Each
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
Adam Equipment LPB 223i Precision Toploading Balance, 220g Capacity x 0.001g Readability, Internal  Calibration EW-11124-72 Mfr # LPB 223I
COMPARE
Internal 220 0.001
$1,036.30USD / Each
$1,205.00USD / Each
Adam Equipment LPB 223e Precision Toploading Balance, 220g Capacity x 0.001g Readability, External Calibration EW-11124-73 Mfr # LPB 223E
COMPARE
External 220 0.001
$877.20USD / Each
$1,020.00USD / Each
Adam Equipment LPB 423i Precision Toploading Balance, 420g Capacity x 0.001g Readability,  Internal  Calibration EW-11124-74 Mfr # LPB 423I
COMPARE
Internal 420 0.001
$1,191.10USD / Each
$1,385.00USD / Each
Adam Equipment LPB 423e Precision Toploading Balance, 420g Capacity x 0.001g Readability,  External Calibration EW-11124-75 Mfr # LPB 423E
COMPARE
External 420 0.001
$1,019.10USD / Each
$1,185.00USD / Each
Adam Equipment LPB 623i Precision Toploading Balance,620g Capacity x 0.001g Readability,  Internal  Calibration EW-11124-76 Mfr # LPB 623I
COMPARE
Internal 620 0.001
$1,380.30USD / Each
$1,605.00USD / Each
Adam Equipment LPB 623e Precision Toploading Balance, 620g Capacity x 0.001g Readability,  External Calibration EW-11124-77 Mfr # LPB 623E
COMPARE
External 620 0.001
$1,199.70USD / Each
$1,395.00USD / Each
Adam Equipment LPB 823i Precision Toploading Balance, 820g Capacity x 0.001g Readability,  Internal  Calibration EW-11124-78 Mfr # LPB 823I
COMPARE
Internal 820 0.001
$1,569.50USD / Each
$1,825.00USD / Each
Adam Equipment LPB 823e Precision Toploading Balance, 820g Capacity x 0.001g Readability,  External Calibration EW-11124-79 Mfr # LPB 823E
COMPARE
External 820 0.001
$1,388.90USD / Each
$1,615.00USD / Each
Adam Equipment LTB 2602i Precision Toploading Balance, 2600g Capacity x 0.01g Readability,  Internal  Calibration EW-11124-80 Mfr # LTB 2602I
COMPARE
Internal 2600 0.01
$1,036.30USD / Each
$1,205.00USD / Each
Adam Equipment LTB 2602e Precision Toploading Balance, 2600g Capacity x 0.01g Readability,  External Calibration EW-11124-81 Mfr # LTB 2602E
COMPARE
External 2600 0.01
$860.00USD / Each
$1,000.00USD / Each
Adam Equipment LTB 3602i Precision Toploading Balance, 3600g Capacity x 0.01g Readability,  Internal  Calibration EW-11124-82 Mfr # LTB 3602I
COMPARE
Internal 3600 0.01
$1,135.20USD / Each
$1,320.00USD / Each
Adam Equipment LTB 3602e Precision Toploading Balance, 3600g Capacity x 0.01g Readability,  External Calibration EW-11124-83 Mfr # LTB 3602E
COMPARE
External 3600 0.01
$946.00USD / Each
$1,100.00USD / Each
Adam Equipment LTB 4602i Precision Toploading Balance, 4600g Capacity x 0.01g Readability,  Internal  Calibration EW-11124-84 Mfr # LTB 4602I
COMPARE
Internal 4600 0.01
$1,225.50USD / Each
$1,425.00USD / Each
Adam Equipment LTB 4602e Precision Toploading Balance, 4600g Capacity x 0.01g Readability,  External Calibration EW-11124-85 Mfr # LTB 4602E
COMPARE
External 4600 0.01
$1,057.80USD / Each
$1,230.00USD / Each
Adam Equipment LBB 6001e Precision Toploading Balance, 6000g Capacity x 0.1g Readability,  External Calibration EW-11124-86 Mfr # LBB 6001E
COMPARE
External 6000 0.1
$1,010.50USD / Each
$1,175.00USD / Each
Adam Equipment LTB 6002i Precision Toploading Balance, 6000g Capacity x 0.01g Readability,  Internal  Calibration EW-11124-87 Mfr # LTB 6002I
COMPARE
Internal 6000 0.01
$1,844.70USD / Each
$2,145.00USD / Each
Adam Equipment LTB 6002e Precision Toploading Balance, 6000g Capacity x 0.01g Readability,  External Calibration EW-11124-88 Mfr # LTB 6002E
COMPARE
External 6000 0.01
$1,659.80USD / Each
$1,930.00USD / Each
Adam Equipment LBB 8001e Precision Toploading Balance, 8000g Capacity x 0.1g Readability,  External Calibration EW-11124-89 Mfr # LBB 8001E
COMPARE
External 8000 0.1
$1,100.80USD / Each
$1,280.00USD / Each
Adam Equipment LBB 12001e Precision Toploading Balance, 12000g Capacity x 0.1g Readability,  External Calibration EW-11124-90 Mfr # LBB 12001E
COMPARE
External 12000 0.1
$1,496.40USD / Each
$1,740.00USD / Each
Adam Equipment LBB 15001e Precision Toploading Balance, 15000g Capacity x 0.1g Readability,  External Calibration EW-11124-91 Mfr # LBB 15001E
COMPARE
External 15000 0.1
$1,685.60USD / Each
$1,960.00USD / Each
Showing 1- 20 of 20