Adam Equipment Luna Analytical Balances
Color-coded navigation buttons facilitate quick recognition
  • Level indicator and adjustable feet ensure proper balance setup
  • Sealed keypad protects against dirt and spills
  • USB and RS-232 interfaces provide fast communication with computer/printer
  • Multilingual display permits use in various regions
  • Selectable digital filtering for animal/dynamic weighing
  • Security slot for optional Kensington-type lock and cable to prevent theft
  • Auto power-off saves energy
Designed with an array of features and functions, the Luna analytical balances accommodate most lab applications to provide faster, more accurate readings. Functions include weighing, parts counting, percentage weighing, dynamic/animal weighing, density determination, checkweighing, and net total weighing. A backlit LCD with 24-mm digits in blue on a black background makes it is easy for you to read in any lighting conditions. Simple to operate, the navigation buttons are color-coded allowing you to perform functions with minimal training.
USB and RS-232 interfaces facilitate data collection and transmission to any computer or printer. GLP printouts are available with time, date and other essential information. Constructed of ABS plastic with a rugged 304 stainless steel pan, these balances are easy to clean and able to withstand demanding lab use. The balances offer a readability of 0.0001 g and can perform basic and detailed measuring tasks.
ITEM(S) ON SALE
$1,371.70 - $2,240.30USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
Adam Equipment LAB 84i  Analytical Balance, 80g Capacity x 0.0001g Readability, Internal Calibration EW-11124-64 Mfr # LAB 84I
COMPARE
Internal 80 0.0001
$1,569.50USD / Each
$1,825.00USD / Each
Adam Equipment LAB 84e  Analytical Balance, 80g Capacity x 0.0001g Readability, External Calibration EW-11124-65 Mfr # LAB 84E
COMPARE
External 80 0.0001
$1,371.70USD / Each
$1,595.00USD / Each
Adam Equipment LAB 124i  Analytical Balance, 120g Capacity x 0.0001g Readability, Internal Calibration EW-11124-66 Mfr # LAB 124I
COMPARE
Internal 120 0.0001
$1,857.60USD / Each
$2,160.00USD / Each
Adam Equipment LAB 124e Analytical Balance, 120g Capacity x 0.0001g Readability, External Calibration EW-11124-67 Mfr # LAB 124E
COMPARE
External 120 0.0001
$1,569.50USD / Each
$1,825.00USD / Each
Adam Equipment LAB 254i  Analytical Balance,250g Capacity x 0.0001g Readability, Internal Calibration EW-11124-68 Mfr # LAB 254I
COMPARE
Internal 250 0.0001
$2,240.30USD / Each
$2,605.00USD / Each
Adam Equipment LAB 254e  Analytical Balance, 250g Capacity x 0.0001g Readability, External Calibration EW-11124-69 Mfr # LAB 254E
COMPARE
External 250 0.0001
$2,055.40USD / Each
$2,390.00USD / Each
Adam Equipment LAB 214i  Analytical Balance, 210g Capacity x 0.0001g Readability, Internal Calibration EW-11124-70 Mfr # LAB 214I
COMPARE
Internal 210 0.0001
$1,952.20USD / Each
$2,270.00USD / Each
Adam Equipment LAB 214e  Analytical Balance, 210g Capacity x 0.0001g Readability, External Calibration EW-11124-71 Mfr # LAB 214E
COMPARE
External 210 0.0001
$1,767.30USD / Each
$2,055.00USD / Each
Showing 1- 8 of 8