Adam Equipment GBK Industrial Scales
Built tough to meet a variety of applications
  • Battery and AC operation
  • Full numeric keypad lets you preset tare, select sample size (count mode), and lock out functions to prevent unauthorized tampering with scale settings
  • Tilt or rotate the adjustable pillar-mounted indicator to suit your viewing needs
  • Large backlit LCD with capacity tracker helps prevent overload
  • Parts counting with selectable sample size
  • Checkweighing LED lights show acceptable limits
  • Removable stainless steel pan for easy cleaning
  • Automatic accumulation for adding totals
  • Bidirectional RS-232 output
  • Capacities up to 120 kg/260 lb
Built tough for a variety of industries the GBK can meet the demands of the warehouse or the production floor. Check weighing applications are trouble free with amber, green, and red indicators to show the operator if the item is under, over, or at the preset limits.
ITEM(S) ON SALE
$692.30 - $808.40USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
Adam Equipment GBK 16A Industrial Scale, 16Lb/8kg Capacity and 0.0002Lb/0.1g Readability Universal Voltage EW-11119-79 Mfr # GBK 16A
COMPARE
8 16 115 / 220
$808.40USD / Each
$940.00USD / Each
Adam Equipment GBK 35A Industrial Scale, 35Lb/16kg Capacity and 0.001Lb/0.5G Readability Universal Voltage EW-11119-81 Mfr # GBK 35A
COMPARE
16 35 115 / 220
$692.30USD / Each
$805.00USD / Each
Adam Equipment GBK 70A Industrial Scale, 70lb/32kg Capacity and 0.002Lb/1g Readability Universal Voltage EW-11119-83 Mfr # GBK 70A
COMPARE
32 70 115 / 220
$752.50USD / Each
$875.00USD / Each
Adam Equipment GBK 130A Industrial Scale, 130lb/60kg Capacity and 0.005Lb/2g Readability Universal Voltage EW-11119-85 Mfr # GBK 130A
COMPARE
60 130 115 / 220
$752.50USD / Each
$875.00USD / Each