Adam Equipment Core Portable Balances
Everything you need for basic weighing
  • Backlit display offers clear view of weighing results at a glance
  • Built-in security slot accommodates an optional Kensington-type lock and cable, deterring theft
  • Auto power-off conserves battery power
  • Vivid backlit LCD easily visible in any lighting conditions
  • Three-point ShockProtect® feature helps prevent overload damage to internal components
  • Removable draft shield on models up to 1500g provides greater weighing accuracy
No other balance can beat the Core for value and ease of use. Rugged ABS housing withstands field environments, and the stainless steel pan removes for easy cleaning. The integrated three-point overload ShockProtect feature safeguards the load cell. The Core operates by 6x AA batteries or AC adapter (cUL certified), while auto power-off saves battery life. A convenient weigh-below hook facilitates below-balance weighing for density and specific gravity measurements.
ITEM(S) ON SALE
$180.60 - $344.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
Adam Equipment CQT202 Core Portable Balance, 200g x 0.01g EW-11712-00 Mfr # CQT 202
COMPARE
200 115 0.01
$296.70USD / Each
$345.00USD / Each
Adam Equipment CQT251 Core Portable Balance, 250g x 0.1g (Lcd) EW-11712-01 Mfr # CQT 251
COMPARE
250 115 0.1
$180.60USD / Each
$210.00USD / Each
Adam Equipment CQT601 Core Portable Balance, 600g x 0.1g (Lcd) EW-11712-02 Mfr # CQT 601
COMPARE
600 115 0.1
$249.40USD / Each
$290.00USD / Each
Adam Equipment CQT1501 Core Portable Balance, 1500g x 0.1g EW-11712-03 Mfr # CQT 1501
COMPARE
1500 115 0.1
$305.30USD / Each
$355.00USD / Each
Adam Equipment CQT2601 Core Portable Balance, 2600g x 0.1g EW-11712-04 Mfr # CQT 2601
COMPARE
2600 115 0.1
$344.00USD / Each
$400.00USD / Each
Adam Equipment CQT5000 Core Portable Balances; 5000g x 1g, 120V EW-11712-06 Mfr # CQT 5000
COMPARE
5000 115 1
$236.50USD / Each
$275.00USD / Each
Adam Equipment CQT202 Core Portable Balance, 200g x 0.01g, 220 V EW-11712-07 Mfr # CQT202_220VAC
COMPARE
200 230 0.01
$296.70USD / Each
$345.00USD / Each
Adam Equipment CQT251 Core Portable Balance, 250g x 0.1g, 220 V EW-11712-08 Mfr # CQT251_220VAC
COMPARE
250 230 0.1
$180.60USD / Each
$210.00USD / Each
Adam Equipment CQT601 Core Portable Balance, 600g x 0.1g, 220 V EW-11712-09 Mfr # CQT601_220VAC
COMPARE
600 230 0.1
$249.40USD / Each
$290.00USD / Each
Adam Equipment CQT5000 Core Portable Balance, 5000g x 1g, 220 V EW-11712-13 Mfr # CQT 5000-220V
COMPARE
5000 230 1
$236.50USD / Each
$275.00USD / Each
Showing 1- 10 of 10