Adam Equipment AGB Industrial Bench Scales
Rugged construction withstands demanding industrial applications
  • Large backlit LCD makes for easy viewing in any lighting conditions
  • Color-coded keys provide quick recognition of frequently used functions
  • Bidirectional RS-232 interface offers quick connection to computers and printers
  • External calibration allows for verification and adjustment with weights
  • Heavy-duty rubber feet ensure stability
  • Stainless steel platform for easy cleaning
  • Programmable auto power-off saves energy
The AGB bench weighing scales are designed to withstand the daily demands in an industrial environment. These scales feature an IP67-rated stainless steel indicator, a large stainless steel weighing platform, and heavy-duty rubber feet to ensure stability. The large backlit LCD with 1½” (40 mm) high digits makes viewing results easy in any lighting condition and changes color for quick notification of checkweighing limits. The easy-to-operate color-coded keypad offers smooth operation and the bidirectional RS-232 interface provides quick connection to computers and printers.
Other features include display hold, audible alarm for checkweighing, capacity tracker to monitor overloads, automatic zero tracking, parts counting with preset sample sizes, preset tare function, selectable digital filtering for animal/dynamic weighting, accumulation function that stores and recalls individual weights or counts, and real-time clock. A built-in rechargeable battery with low battery indicator provides up to 90 hours of operation.
ITEM(S) ON SALE
$877.20 - $950.30USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Readability (g) Description
Availability Pricing
Adam Equipment AGB 175a Industrial Bench Scale, 175lb/80kg x 0.005lb/2g, 11.8"x15.7" Platform EW-11119-60 Mfr # AGB 175A
COMPARE
80 2 Industrial Bench Scale, 175lb/80kg x 0.005lb/2g, 11.8\"x15.7\" Platform
$950.30USD / Each
$1,105.00USD / Each
Adam Equipment AGB 35a Industrial Bench Scale, 35lb/16kg x 0.001lb/0.5g, 11.8"x15.7" Platform EW-11119-64 Mfr # AGB 35A
COMPARE
16 0.5 Industrial Bench Scale, 35lb/16kg x 0.001lb/0.5g, 11.8\"x15.7\" Platform
$877.20USD / Each
$1,020.00USD / Each
Adam Equipment AGB 65a Industrial Bench Scale, 65lb/30kg x 0.002lb/1g, 11.8"x15.7" Platform EW-11119-65 Mfr # AGB 65A
COMPARE
30 1 Industrial Bench Scale, 65lb/30kg x 0.002lb/1g, 11.8\"x15.7\" Platform
$941.70USD / Each
$1,095.00USD / Each