This product is now discontinued, View Related Products
Accessories for Koehler Fluorescent Indicator Adsorption Apparatus
Optimize performance of your Koehler Fluorescent Indicator Adsorption Apparatus
Compatible for use with the Koehler Fluorescent Indicator Absorption Apparatus, sold separately.
An integrated electric vibration system facilitates the dry gel packing of the columns.
After the sample is introduced and adsorbed into the column, fluorescent dye indicators mark the boundaries of aromatics, olefins, and saturates, which are visible with a UV lamp.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Koehler K41500-4 Fluorescent Indicator Apparatus Silica Gel; 500 gram EW-59876-87 Mfr # K41500-4
COMPARE
Fluorescent Indicator Apparatus Silica Gel; 500 gram
Call For Price
Koehler K41500-5 Fluorescent Indicator Apparatus Dried Silica Gel; 40 gram EW-59876-88 Mfr # K41500-5
COMPARE
Fluorescent Indicator Apparatus Dried Silica Gel; 40 gram