A&D Weighing FC-Si and FC-i Counting Scales
Automatic Counting Accuracy Improvement (ACAI) ensures high counting accuracy
  • Store/retrieve 500 unit weights by ID numbers
  • LED prompting navigation makes it easy to use
The FCi Series counting scales utilize ACAI, A&D’s exclusive Automatic Count Accuracy Improvement. ACAI combines the two most critical performance requirements of a counting scale: count accuracy & operation expediency. Initial Average Piece Weight can be determined through an operator’s choice of fixed, random, keyboard entry, or memory recall.
ITEM(S) ON SALE
$1,245.00 - $1,684.90USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (lbs) Power (VAC) Readability (g)
Availability Pricing
A&D Weighing FC-5000Si Hi-Res Counting Scale 5kg x 0.2g / 10lb x 0.0005Lb 500 Memory Sets EW-11110-49 Mfr # FC-5000SI
COMPARE
10 115 0.2
$1,684.90USD / Each
$2,030.00USD / Each
A&D Weighing FC-500SI Counting Scale, 500g, 115 V/60 Hz EW-11113-05 Mfr # FC-500SI
COMPARE
1 115 0.02
$1,684.90USD / Each
$2,030.00USD / Each
A&D Weighing FC-5000I Counting Scale, 5000g, 115 V/60 Hz EW-11113-09 Mfr # FC-5000I
COMPARE
10 115 0.5
$1,245.00USD / Each
$1,500.00USD / Each
A&D Weighing FC-10KI Counting Scale, 10, 000g Capacity EW-11113-11 Mfr # FC-10KI
COMPARE
20 115 1
$1,245.00USD / Each
$1,500.00USD / Each
A&D Weighing FC-20KI Counting Scale, 20, 000g Capacity EW-11113-13 Mfr # FC-20KI
COMPARE
50 115 2
$1,245.00USD / Each
$1,500.00USD / Each
A&D Weighing FC-50KI Counting Scale, 50, 000g Capacity EW-11113-15 Mfr # FC-50KI
COMPARE
100 115 5
$1,245.00USD / Each
$1,500.00USD / Each
A&D Weighing FC-2000i Hi-Resolution 5Lb Counting Scale EW-11115-79 Mfr # FC-2000I
COMPARE
5 115 0.2
$1,245.00USD / Each
$1,500.00USD / Each
A&D Weighing FC-500i Hi-Resolution 1Lb Counting Scale EW-11115-80 Mfr # FC-500I
COMPARE
1 115 0.05
$1,245.00USD / Each
$1,500.00USD / Each
Showing 1- 8 of 8