A&D EK-L Compact Bench Scales
Greater precision for large samples
  • High-resolution display combines with large capacity for critical bulky, dense, or heavy-tare applications
  • Durable 430 SS weighing pan accommodates oddly shaped and oversized items
  • Large backlit LCD is viewable in any lighting conditions
  • Output data to PC or printer via bi-directional RS-232 interface
  • Comparator function provides fast accurate filling, packing, and sorting
  • Use a cable lock with the built-in security ring to prevent theft
  • Dual-range 30 kg model automatically switches from low to high when the weight exceeds 15 kg
Choose the bench scale with the capacity you need.
ITEM(S) ON SALE
$891.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
A&D Weighing EK-L Compact Bench Scale, 15 kg x 0.1 g EW-11133-07 Mfr # EK-15KL
COMPARE
Compact Bench Scale, 15 kg x 0.1 g
$891.00USD / Each
$990.00USD / Each
A&D Weighing EK-L Dual Range Compact Bench Scale, 3 kg x 0.1 g, 30 kg x 1 g EW-11133-09 Mfr # EK-30KL
COMPARE
Dual Range Compact Bench Scale, 3 kg x 0.1 g, 30 kg x 1 g
$891.00USD / Each
$990.00USD / Each