6-Watt, Short/Long-Wave Ultraviolet Lamp
Get short and long-wave ultraviolet light at the touch of a button. Two separate tubes (254 and 365 nm) operate independently—no wavelength mixing occurs.
Snap-on filter frame simplifies filter replacement. Filter measures 6" x 1-3/4" and provides high-intensity fluorescence for a minimum of 5000 hours. Wavelength rocker switch is conveniently located for index-finger operation. Housing is constructed of lightweight CYCOLAC(R) plastic. Lamp includes a 7-ft, two-wire cord (115 VAC model has a plug). Order optional lamp stand 09818-50 separately.
$686.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (Hz) Power (VAC) Wavelength
Availability Pricing
Analytik Jena Short/long-wave UV lamp; 6 watts, 254/365 nm wavelength, 115 VAC/60 Hz EW-09818-00 Mfr # 95-0007-05
COMPARE
60 115 254/365
$686.00USD / Each
Analytik Jena Short/long-wave UV lamp; 6 watts, 254/365 nm wavelength, 230 VAC/50 Hz EW-09818-02 Mfr # 95-0007-06
COMPARE
50 230 254/365
$686.00USD / Each