3M Qualitative Fit Test Kits
Meets OSHA requirements
  • Easy to use
  • Provides a consistent environment for testing fit
These fit test kits test the face-to-respirator seal of particulate respirators and gas/vapor respirators with a particulate prefilter. Easy-to-use kits provide a consistent environment for testing fit, leading to reliable results meeting OSHA requirements.
Testing is easy: a sensitivity test is performed first by spraying a dilute version of the test solution into the hood without a respirator being worn to make sure the person can detect the taste. Then, while wearing a respirator, a sweet or bitter test solution is sprayed into the hood. If the wearer doesn't detect the taste of the test agent, the respirator fit is acceptable. If the taste is detected, a second test is performed after readjusting the respirator. If that test also fails, it may indicate the need for a different size or model of respirator.
$387.20USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
3M FT-10 Respirator Qualitative Fit Test Kit, Sweet taste EW-86332-30 Mfr # FT-10
COMPARE
Respirator Qualitative Fit Test Kit, Sweet taste
$387.20USD / Each
3M FT-30 Respirator Qualitative Fit Test Kit, Bitter taste EW-86332-31 Mfr # FT-30
COMPARE
Respirator Qualitative Fit Test Kit, Bitter taste
$387.20USD / Each