Labconco 10- and 32-Pin Rack Insert for SteamScrubber
  • Configure your washer with optional inserts
  • Pins support flasks, beakers, bottles, and other dishes in an upright position
Customize your washer for your labware needs. The SteamScrubber labware washer has an upper and lower rack; however, the rack is undivided. Use these inserts to create areas for smaller labware items, especially those with narrow openings that need to be upright for thorough cleaning.
$236.00 - $330.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Labconco 10-Pin Rack Insert for SteamScrubber EW-98962-66 Mfr # 4401501
COMPARE
10-pin rack insert for SteamScrubber
$236.00USD / Each
Labconco 32-Pin Rack Insert for SteamScrubber EW-98962-68 Mfr # 4401801
COMPARE
32-pin rack insert for SteamScrubber
$330.00USD / Each