Masterflex Single-Use Components
With Masteflex Single-Use, you will no longer have to validate the cleaning and sterilizing of your bioprocess assemblies. The purchase of a preassembled, sterilized, and validated system shifts the principal burden of validation to the supplier. That is why it is important to be sure your supplier uses only the most reliable brand name products and adheres to strict, well-established protocols in the sterilization process.
Masterflex Single-Use assemblies are manufactured in a Class 10,000 cleanroom. The assembly facility is ISO-13485 certified, cGMP compliant and FDA registered. We offer gamma irradiation sterilization with a 10–6 SAL (Sterility Assurance Level), a one millionth probability of microbial survival an full lot traceability that identifies the production site, manufacturing date and origin of the components. We also provide and maintain all required documentation, including complete validation documents and certificates of conformance and sterility.
We build your custom assemblies with reliable brand-name products
It's time to make your process more efficient
  • 1Exactly what you need
    Choose from thousands of components. We custom configure, validate, and package according to your specifications.
  • 2Exactly when you need it
    Stop cleaning and revalidating your system. We manage your inventory and send assemblies according to your schedule.
  • 3Call for a Quote today
    Masterflex Single-Use experts provide a quote with full schematic, followed by an actual sample for your approval