Jenway Spectrophotometers: Specs & Features Comparison
Spectrophotometer Specifications Comparison Chart
Jenway high-quality, easy-to-use, UV/visible spectrophotometers (some models with CPLive™ cloud connectivity) are designed to suit many budgets, industries and applications. Choose from entry-level single beam spectrophotometers, split-beam spectrophotometers, diode array scanning spectrophotometers, and micro-volume spectrophotometers with bandwidths ranging from 8 nm to 1.5 mn. Spectrophotometer equipped with CPLive connectivity provide secure data storage and sharing via the cloud. Refer to the table below to select a Jenway spectrophotometer to meet your requirements.
Table scrolls horizontally
Model Visible UV Scanning Single beam Split beam Double beam Micro-volume Bandwidth
Jenway 632621 Visible Spectrophotometer +     +       8nm
Jenway 6300 Benchtop Visible Spectrophotometer +     +       8nm
Jenway 7200 Visible Diode Array Spectrophotometer +   + +       7nm
Jenway 7205 UV-Vis Diode Array Spectrophotometer + + + +       5nm
Jenway 7410 Cloud-based Visible Spectrophotometer +   + +       5nm
Jenway 7415 Cloud-based UV- Vis Spectrophotometer + + + +       5nm
Jenway 7475 Nano Micro-Volume Spectrophotometer + + + +     + 5nm
7615 (Coming Soon) + + +   +     1.5nm