Zefon Cassette, Air-O-Cell; 10/Bx
Representative image only.
Zefon Item # EW-60061-27
Specifically designed for fast collection of airborne aerosols and inorganic particulates
  • No need to prepare slides or worry about contamination
  • Special adhesive on collection media prevents collected sample from being washed off
  • Attain direct quantitative analysis of organic and inorganic particulate
  • 10/BX
PRODUCT OPTIONS
$52.00USD / Box of 10
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionCassettes, Air-O-Cell®; 10/BX
Key Features
  • No need to prepare slides or worry about contamination
  • Special adhesive on collection media prevents collected sample from being washed off
  • Attain direct quantitative analysis of organic and inorganic particulate
  • 10/BX
More About this Item
Use these disposable, easy-to-use air sampling cassettes to collect a wide range of airborne particulates such as mold, spores, pollen, insect parts, fibers, skin cells, and more. Air-O-Cell cassettes provide a broader overview of potential contaminants than conventional sampling methods by collecting both viable and nonviable sample specimens.
Connect to an air sampling pump to draw particulate-laden sample through the tapered inlet towards a slide. Slide features special collection media with adhesive to prevent particulate from blurring or being washed off during the staining process. Once the sampling is complete, send the Air-O-Cell cassettes to a laboratory, where the slides are removed and analyzed preventing contamination worries.
Air-O-Cell cassettes have a limited shelf life.
Use cassettes with sampling pump capable of drawing 15 L/min (open flow).
Air-O-Cell cassettes have a limited shelf life.