YSI 6051030-20 Pro Series pH/ISE/ORP/Conductivity Cable; 20-meter L
Representative image only.
YSI Mfr # 6051030-20 – Item # EW-59352-76
Optimize the performance of YSI meters
  • Choose from a variety of accessories
  • 6051030 Pro-series cable, 20 meter; for pH, ISE, ORP, and conductivity
  • 2 year warranty
PRODUCT OPTIONS
$798.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithYSI Professional Plus
  • DescriptionPro Series pH/ISE/ORP/Conductivity Cable; 20-meter L
Two year warranty
Key Features
  • Choose from a variety of accessories
  • 6051030 Pro-series cable, 20 meter; for pH, ISE, ORP, and conductivity
  • 2 year warranty
More About this Item
Replacement electrodes, cables, power adapters, and other accessories for your YSI water analysis meters.
Choose the correct cable length to meet your requirements. All cables include a temperature probe and a four cell conductivity probe.