Wilmad-LabGlass High-Throughput NMR Tube and Cap, 4.94665 mm OD x 7" L; 100/Pk
Representative image only.
Wilmad-LabGlass Mfr # WG-1000-7 – Item # EW-83999-58
High-throughput NMR tubes have one of the best OD tolerances in the industry
  • NMR tubes are constructed of ASTM E438 Type 1 Class B borosilicate glass
  • Ensure spectrum clarity with an averaged camber of 60 micron
  • NMR tube features a 7" length, 4.94665 ± 0.01905 mm OD, and 0.43 mm wall thickness
PRODUCT OPTIONS
$151.00USD / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Tube MaterialGlass
  • Tube ColorClear
  • Tube Bottom ShapeRound
  • Tube Length (mm)178
  • Tube Outside Diameter (OD) (mm)4.94665
  • Cap IncludedYes
  • Cap StyleDisposable
  • GraduationsNo
  • Special Features600 MHz
  • DescriptionHigh-Throughput NMR Tube and Cap, 4.94665 mm OD x 7\" L; 100/Pk
Made in the U.S.A.
Includes
NMR tubes and caps.
Key Features
  • NMR tubes are constructed of ASTM E438 Type 1 Class B borosilicate glass
  • Ensure spectrum clarity with an averaged camber of 60 micron
  • NMR tube features a 7" length, 4.94665 ± 0.01905 mm OD, and 0.43 mm wall thickness
More About this Item
These Wilmad-LabGlass high-throughput NMR tubes have an averaged camber of 60 micron to guarantee the spectrum resolution for small molecules (MW<250) samples up to 600 MHz. Each 5mm tube meets precise wall thickness to ensure they fit tightly in spinner turbine. All tubes are made from borosilicate glass. Choose from tubes with authentic Bruker® SampleJet® caps without code or tubes with disposable caps.