Wika A-AI.1 Attachable Loop-Powered Digital Pressure Indicator
Representative image only.
Wika Mfr # 7082534 – Item # EW-68447-13
$393.30USD / Each
sign in for your price.
$175.00USD / Each
for an additional
$175.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Enclosure RatingIP65
  • DescriptionAttachable Loop-Powered Digital Pressure Indicator
More About this Item
  • Attachable liquid crystal display for local indication
  • 4-20 mA current loop-powered with 3 volt drop
  • Installs between transmitter and DIN 43 650, + full sized
    "L" shaped 4-pin connector
  • Programmable Range: 4-20 mA can be assigned any
    display value within the display range
  • User scalable to display range from -1999 to +9999
  • Accuracy: = 0.2% of span