Wearwell 24/Seven® Modular Industrial Anti-Fatigue Mat, Nitrile Rubber, Drainage with GRITSHIELD™, Black, 3 ft x 3 ft

Representative image only.
Wearwell Mfr # 576 58x3x3NBRBK – Item # EW-81853-15
Modular anti-fatigue, anti-slip mats keep employees safe and comfortable
  • Keep workers productive, safe, and comfortable all day, every day
  • Extremely versatile — mats interlock on all sides to cover entire floor areas, long assembly lines, or stand alone as single workstation mats
  • Tough enough to handle 24/7 shift operations
  • For extra safety, attach optional 2" x 39" yellow safety edges
  • Ideal for use with petroleum-based fluids
  • Drainage style mates with perforated 15/16" hold maximizes drainage in wet workspaces
  • GRITSHIELD™ increases traction
  • 1 year warranty

PRODUCT OPTIONS

$224.75USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • MaterialNitrile
  • Thickness (in)0.625
  • Thickness (mm)16
  • Width (in)36
  • Length (in)36
  • Width (cm)91
  • Length (cm)91
  • ColorBlack
  • DescriptionModular Industrial Anti-Fatigue Mat, Nitrile Rubber, Drainage with GRITSHIELD™, Black, 3 ft x 3 ft
  • Warranty1 year
1 Year Warranty

Key Features

  • Keep workers productive, safe, and comfortable all day, every day
  • Extremely versatile — mats interlock on all sides to cover entire floor areas, long assembly lines, or stand alone as single workstation mats
  • Tough enough to handle 24/7 shift operations
  • For extra safety, attach optional 2" x 39" yellow safety edges
  • Ideal for use with petroleum-based fluids
  • Drainage style mates with perforated 15/16" hold maximizes drainage in wet workspaces
  • GRITSHIELD™ increases traction
  • 1 year warranty

More About this Item

The 24/Seven® modular mat provides enhanced comfort and safety with anti-fatigue and cushioned support. Customize matting to your desired size using these 3 ft x 3 ft interlocking modules. The hidden interlocking system is designed to connect on all four sides to quickly cover large areas. Yellow beveled edging provides protection from tripping and a clean finished look; order separately.

Choose from three different rubber compounds based on environmental exposures.
Grease resistant model GR mats are suitable for water or dry debris.
Cutting-fluid resistant model CFR mats are ideal for mineral oil-based fluids.
Nitrile rubber model NBR mats stand up to petroleum-based fluids.

GRITSHIELD™ provides added traction where slip hazards are present. Solid design mats are for dry applications and open drainage models have large holes to allow fluid and debris to flow through.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat