W.S. Tyler 8576 Round Sieve Brush for 100 Mesh Sieves and Coarser, Brass Bristles
Representative image only.
W.S. Tyler Mfr # 8576 – Item # EW-59986-97
Keeps sieves clean for better separation
  • Will not damage or hinder performance of sieve
  • 3 month warranty
PRODUCT OPTIONS
$51.80USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Length (in)6
  • Diameter (in)2
  • DescriptionRound Sieve Brush for 100 Mesh Sieves and Coarser, Brass Bristles
  • Warranty3 Months
3 Month Warranty
Key Features
  • Will not damage or hinder performance of sieve
  • 3 month warranty
More About this Item
These round sieve brushes are ideal for cleaning fine and coarse mesh sieves. The nylon brush is to clean 120 mesh and finer while brass bristle brushes are more suited for 100 mesh and coarser. Proper maintenance and cleaning of sieves prevents damage to sieve openings, keeping them clean and free of particles and ensures longest possible in-spec service life and accurate results.
Each brush measures 6” long and the diameter of the bristle head is 2”.
Use this brush with coarse mesh sieves, 100 mesh and coarser. Brush has brass bristles and wood handle.