Troemner WA229 8" S/S Tweezer Smooth Tip for mg, Weights
Representative image only.
Troemner Mfr # WA229 – Item # EW-01027-95
SPECS & DESCRIPTION
Specifications & Description
  • Overall Length (in)8
  • Overall Length (cm)20.32
  • MaterialStainless steel
  • DescriptionTroemner WA229 8\" S/S Tweezer Smooth Tip for mg, Weights
More About this Item
Forceps / Tweezers are designed for proper handling of weights.
Stainless steel forceps are available in 5” and 8” sizes and are designed in a smooth tip, curved tip, straight grip tip, and a 22° grip tip. Stainless steel forceps tips are made of PVC.
Economical polycarbonate forceps for weights 1 mg through 200 g.