Troemner WA228 8" S/S Tweezer Grip Tip for Weights 1g Through 200g

Representative image only.
Troemner Mfr # WA228 – Item # EW-01027-94

$92.50USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Overall Length (in)8
  • Overall Length (cm)20.32
  • MaterialStainless steel
  • DescriptionTroemner WA228 8\" S/S Tweezer Grip Tip for Weights 1g Through 200g

More About this Item

Forceps / Tweezers are designed for proper handling of weights.

Stainless steel forceps are available in 5” and 8” sizes and are designed in a smooth tip, curved tip, straight grip tip, and a 22° grip tip. Stainless steel forceps tips are made of PVC.

Economical polycarbonate forceps for weights 1 mg through 200 g.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat