Troemner 9498 50 lb, 10-1/4" Dia, Avoirdupois Slotted Weights
Representative image only.
Troemner Mfr # 9498 – Item # EW-11966-59
Use in a variety of applications such as pressure, torque, and tensile strength testing
  • Use several slotted weights together to build up from a minimum weight to a maximum test load
  • A durable coat of paint protects each weight from rusting and chipping
  • Color coded – silver for avoirdupois
  • 3 month warranty
PRODUCT OPTIONS
$303.00USD / Each
$112.00 - $185.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • Mass ClassClass 6
  • Mass Size (lbs)50
  • Mass CertificationNone
  • Thickness (in)2.75
  • Diameter (in)10.25
  • Slot0.5625\"
  • Description50 lb, 10-1/4\" Dia, Avoirdupois Slotted Weights
  • Warranty3 month
3 Month Warranty
Key Features
  • Use several slotted weights together to build up from a minimum weight to a maximum test load
  • A durable coat of paint protects each weight from rusting and chipping
  • Color coded – silver for avoirdupois
  • 3 month warranty
More About this Item
Cast Iron Slotted Weights are used in a variety of applications such as pressure, torque, and tensile strength testing. Cast Iron Slotted Weights are typically used with a hanger that also has its weight calibrated so the hanger can be used as part of the overall weight under test. Several Cast Iron Slotted Weights may be used together to build up from a minimum weight to a maximum test load. The hanger weight selected should be able to accommodate the total load needed.
Cast Iron Slotted Weights above 2 kg/5 lb have adjusting cavities. Weights below 2 kg/5 lb are adjusted by removing material from the underside of the weight. Each weight has its nominal value cast into the topside of the weight.