Traceable 6412 Scientific Single-Input RTD Thermometer with USB and Probe

Representative image only.
Traceable Item # EW-37804-27
A highly accurate thermometer that is simple to use
  • NIST-traceable certificate
  • Traceable® model number 6412
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Meter Measurement TypeRTD
  • Number Of Probe Channels1
  • Types Of Probes AcceptedRTD
  • Min Temperature (° F)-328
  • Max Temperature (° F)932
  • Min Temperature (° C)-200
  • Max Temperature (° C)500
  • Accuracy±0.05°C between 0° to 100°C
  • Resolution0.001°
  • Display Type0.4\" Display, 6-digit LCD
  • Height (in)5 1/2
  • Depth (in)3 1/2
  • No. of Probe(s) Included1
  • Probe Diameter (in)0.125
  • Probe Length (in)6 21/64
  • Battery Life1 year
  • Battery9v Battery (1)
  • DescriptionScientific Single-Input RTD Thermometer with USB and Probe
  • Warranty1 Year
1 Year WarrantyConformite Europeenne

Key Features

  • NIST-traceable certificate
  • Traceable® model number 6412
  • 1 year warranty

More About this Item

This high-precision thermometer is ideal for measuring temperatures in gas systems, reagents, chemical solutions, soil, water baths, ovens, incubators, pharmaceuticals, foods and wastewater. Designed for years of reliable service even in severe environments, the meter features a splash proof membrane keypad. A high-contrast 1/2"-high LCD makes the thermometer easy to read in all types of light. The meter features memory and hold function, internal datalogger that captures the min/max temperature every hour for a 24-hours period, and °F/°C selection. For easy reading, the meter can be changed to display one, two, or three significant numbers after the decimal point. The fast-response general purpose probe (included) displays even the slightest temperature change instantly. Readings are updated three times a second. The internal memory can be downloaded into a computer using optional software 90080-21.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat