Traceable 4233 Pocket Thermocouple Probe K Thermometer, C
Representative image only.
Traceable Item # EW-90225-20
  • Traceable® model number 4233
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeThermocouple
  • Types Of Probes AcceptedType K
  • DescriptionPocket Thermocouple Probe K Thermometer, C
  • Warranty1 Year
1 Year Warranty
Key Features
  • Traceable® model number 4233
  • 1 year warranty
More About this Item
Shockproof, pocket size thermometer is ideal for lab, plant, and field use
To assure accuracy an individually serial-numbered Traceable® Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST (National Institute of Standards and Technology).
Features:
  • Supplied with Fast-response beaded Type-K probe
  • Size is 2¾ x 4¼ x 7/8 inches
  • Weight is 4 ounces
Guaranteed to have at least one year of calibration remaining on certificate.