Thermo Scientific STAR A214 Star A214 pH/ISE Fluoride Meter Kit

Representative image only.
Thermo Scientific Mfr # STARA2147 – Item # EW-58825-32
Durability and dependability make it the ideal choice for every lab, every sample, every day
  • Multilanguage interface supports English, Spanish, French, Italian, German, and Chinese
  • Improved stability algorithm provide better measurement accuracy
  • Ensure temperature accuracy with ATC temperature calibration
  • Nonvolatile memory holds up to 2000 data points with time date stamp
  • Individual method passwords provide better security
  • Option of AC power or four AA batteries (sold separately)
  • 3 year warranty

PRODUCT OPTIONS

$4,027.50USD / Each
$4,475.00USD / Each

$164.50 - $237.00USD / Each

Time to recalibrate? Click on the calibration service to view details, then order.
Include Electrodes Today!
Include Electrodes Today!

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeBenchtop
  • Display typeGraphic LCD
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.002 pH
  • Buffer RecognitionDIN, NIST, and USA
  • CalibrationUp to 5 points
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1 or 1 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Min ISE0 ppm
  • Max ISE19999 ppm
  • ISE Resolution0.01 ppm
  • ISE Accuracy±0.2 or 0.05% of reading
  • Dissolved Oxygen Accuracy±0.5% of reading
  • Temperature compensationAutomatic or manual
  • WaterproofYes
  • Width (cm)17.78
  • Height (cm)24.765
  • Width (in)7
  • Length (in)3 3/8
  • Height (in)9 3/4
  • DescriptionStar A214 pH/ISE Fluoride Meter Kit
  • Warranty3 year
3 Year WarrantyIP54 RatedISO9001:2000 Certified Supplier

Key Features

  • Multilanguage interface supports English, Spanish, French, Italian, German, and Chinese
  • Improved stability algorithm provide better measurement accuracy
  • Ensure temperature accuracy with ATC temperature calibration
  • Nonvolatile memory holds up to 2000 data points with time date stamp
  • Individual method passwords provide better security
  • Option of AC power or four AA batteries (sold separately)
  • 3 year warranty

More About this Item

Thermo Scientific Orion Star A214 pH, ISE, mV, RmV and temperature meter features an intuitive user interface with plain language screen prompts. A large backlit, graphic LCD shows measurement with temperature along with electrode status, time/date, sample ID, user ID, and calibration data.

Up to five point automatic calibration with USA/NIST and DIN buffers, auto-buffer recognition, and recalibration feature to fix calibration errors. GLP-related features include 2000 data points with time/date stamp and electrode verification. Meter utilizes RS-232 or USB for connectivity and includes data analysis software.

Meter kit includes Fluoride ISE (05723-02), ROSS™ pH electrode (05718-02), Star ATC probe (58824-48), Star stirring probe (58823-45), 475-mL each of 1ppm, 2ppm, 10ppm standards, 1-gallon of TISABII, swing-arm electrode holder, and universal power adapter.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat