Thermo Scientific R060232 Prepared Media Tube, Bhi Agar Slant 20/pk
Representative image only.
Thermo Scientific Mfr # R060232 – Item # EW-14207-40
Provide fast and efficient quality results!
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
SPECS & DESCRIPTION
Specifications & Description
  • Media Name (No.)Brain-Heart Infusion
  • Tube Length (mm)103
  • Tube Diameter (mm)15
  • DescriptionPrepared Media Tube, Bhi Agar Slant 20/pk
Key Features
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
More About this Item
Choose from a variety of media formulations. Use this culture media for the isolation and differentiation of microorganisms from samples or specimens. Most are for immediate use — no further preparation is necessary. Each raw component is pretested before being used, for quality assurance and consistency from lot batch.