Thermo Scientific Nalgene 5030-0510 Cryogenic vial holder for 50 (1.0 to 5.0 mL) vials

Representative image only.
Thermo Scientific Nalgene Mfr # 5030-0510 – Item # EW-06754-90
Polycarbonate vial holders features special design that locks the bottom of the vial in place
  • 1 year warranty

PRODUCT OPTIONS

$68.80USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Tubes Held50
  • Tube Size1.0 to 5.0 mL
  • ColorWhite and Blue
  • Height (in)1
  • Height (cm)2.5
  • DescriptionCryogenic vial holder for 50 (1.0 to 5.0 mL) vials
  • Warranty1 year
1 Year Warranty

Key Features

  • 1 year warranty

More About this Item

Polycarbonate vial holders allow single-hand opening/closing of cryogenic vials. Alphanumeric locating system is molded into the upper surface for easy sample identification. Rubber feet prevent sliding. Autoclavable.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat