Thermo Scientific C-4000-55B Caps with Septa; pre-slit blue PTFE/white silicone
Representative image only.
Thermo Scientific Mfr # 6PSC9STS1 – Item # EW-98810-24
A variety of septa materials to match your equipment and application
  • Prevent septa push through and over- and under-tightening
  • Easy-on, easy-off convenience with just one turn
PRODUCT OPTIONS
$86.50USD / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionCaps with Septa; pre-slit blue PTFE/white silicone
Key Features
  • Prevent septa push through and over- and under-tightening
  • Easy-on, easy-off convenience with just one turn
More About this Item
Get the assurance of a crimp seal with the time-saving convenience of a screw cap. The unique threaded crown design ensures a secure seal with just a half-turn of the cap.
Vials sold separately.