Thermo Scientific 9711 Clearing Agent, Toluene; 4 x 1 gal
Representative image only.
Thermo Scientific Mfr # 9711 – Item # EW-48800-30
Ideal for tissue processing and staining
  • Packaged in high-density polyethylene easy-to-pour container
  • 1 month warranty
$161.25USD / Case of 4
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (L)15.14
  • ColorClear
1 Month Warranty
Key Features
  • Packaged in high-density polyethylene easy-to-pour container
  • 1 month warranty
More About this Item
These clearing solutions are specifically designed for tissue processing and the staining of histological and cytological specimens.
Toluene can be used with both open and closed tissue processors. It is also compatible with all manual staining procedures and automatic stainers. Please note, toluene is a flammable product and must be stored in a flammable fire cabinet. Protective gloves should be worn and work should be performed in a ventilated area.