Thermo Scientific 88871003 Compact Dry Block Heater, Single Block, 100-240V, Eu / Apac Plugs
Representative image only.
Thermo Scientific Mfr # 88871003 – Item # EW-36403-36
Maximize bench space with our compact, lightweight dry bath / block heater
  • Custom block modules provide flexibility for heating different size tubes
  • Built-in temperature sensing probe for outstanding temperature accuracy and control
  • 12V low-voltage power input
  • 2-1/2 Years Parts and Labor Warranty
SPECS & DESCRIPTION
Specifications & Description
  • Plug TypeUS Standard
  • Power (VAC)100 to 240
  • Width (in)7 5/16
  • Length (in)7 5/16
  • Height (in)1
  • DescriptionCompact Dry Block Heater, Single Block, 100-240V, Eu / Apac Plugs
  • Warranty2-1/2 Years Parts and Labor
2 and a Half Year Warranty
Key Features
  • Custom block modules provide flexibility for heating different size tubes
  • Built-in temperature sensing probe for outstanding temperature accuracy and control
  • 12V low-voltage power input
  • 2-1/2 Years Parts and Labor Warranty
More About this Item
Increase lab versatility with the Thermo Scientific compact digital dry baths / block heaters, which offer a wide range of configurations with interchangeable modular blocks for a variety of applications. These heaters are portable and designed with a compact footprint to save valuable bench space. Built-in temperature-sensing probe assists with outstanding temperature accuracy and control. For a complete system, order both a standard or advanced digital controlled heater and the desired heating block modules.