Thermo Scientific 8104BNUWP ROSS Ultra pH Probe, DJ/Glass/Rugged-Bulb; WP BNC
Representative image only.
Thermo Scientific Mfr # 8104BNUWP – Item # EW-05718-73
18 months replacement warranty
  • Builds on the ROSS® technology for greater stability, relability, and renowned accuracy and temperature response
  • Less long term drift saves time from recalibrating
  • Double junction
  • Glass body
  • Rugged-bulb
  • Waterproof BNC connection
  • 1 year warranty
PRODUCT OPTIONS
$1,141.20USD / Each
$1,268.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • Reference JunctionCeramic
  • Reference ElectrolyteROSS
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Temperature CompensationNo
  • ConnectorBNC
  • Cable Length (m)1
  • DescriptionUltra pH Probe, DJ/Glass/Rugged-Bulb; WP BNC
  • Warranty1 year
1 Year Warranty
Key Features
  • Builds on the ROSS® technology for greater stability, relability, and renowned accuracy and temperature response
  • Less long term drift saves time from recalibrating
  • Double junction
  • Glass body
  • Rugged-bulb
  • Waterproof BNC connection
  • 1 year warranty
More About this Item
The ROSS Ultra stability gives you ±0.01 pH accuracy even over a long period of time. Using a nonmetallic reference system, the liquid-liquid equilibration occurs almost instantaneously, saving time plus ensuring accurate results. Complex, delicate samples, such as biological media, foods, and pharmaceuticals, measure easily without metallic contamination of the sample or excessive cleaning. ROSS Ultra electrodes are refillable—easily accommodating a change in filling solution for samples sensitive to potassium or chloride ions. Choose ROSS Ultra pH/ATC Triodes for applications requiring built-in temperature compensation.