Thermo Scientific 8104BN pH Probe, Refillable/DJ/Glass/Rugged-Bulb; BNC
Representative image only.
Thermo Scientific Mfr # 8104BN – Item # EW-05718-27
Drift less than 0.002 pH per day
  • Platinum wire prevents contamination with silver and mercury ions
  • Excellent for use in tris buffer
  • 1 year warranty
PRODUCT OPTIONS
$1,043.10USD / Each
$1,159.00USD / Each
Suggested alternative
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsField use-noaqueous, DI-water
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • Reference JunctionCeramic
  • Reference ElectrolyteROSS
  • Min pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • ConnectorBNC
  • Cable Length (m)1
  • DescriptionpH Probe, Refillable/DJ/Glass/Rugged-Bulb; BNC
  • Warranty1 year
1 Year Warranty
Key Features
  • Platinum wire prevents contamination with silver and mercury ions
  • Excellent for use in tris buffer
  • 1 year warranty
More About this Item
These provide fast, accurate, and stable pH readings. Ideal for food, pharmaceuticals, or biological samples. Double ceramic junction design protects the inner reference solution and prolongs the life of the electrode. Readings stabilize to 0.01 pH in less than 30 seconds—even with a 50degC temperature change. Electrodes read pH from 0 to 14. Max temperature is 90degC for glass electrodes and 80degC for epoxy. All electrodes have BNC connectors.
Models 05718-20 and 05721-26 have a pin-tip connector, which is identical to the pin-tip portion of a U.S. Standard connector.