Tempilaq Temperature-Indicating Paint, 750⁰F (2-oz Jar)
Representative image only.
Tempil Mfr # TEMPLAQ0750F2OZ – Item # EW-09036-80
Easy confirmation that a target temperature has been met
  • Nonflammable for maximum safety
  • Easy application with quick-dry formula on any surface
  • 1 year warranty
PRODUCT OPTIONS
$46.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Number of Temperature Indicators1
  • Temperature Points (° F)750
  • Temperature Points (° C)399
  • DescriptionTemperature-Indicating Paint, 750⁰F (2-oz Jar)
  • Warranty1 year
1 Year Warranty
Key Features
  • Nonflammable for maximum safety
  • Easy application with quick-dry formula on any surface
  • 1 year warranty
More About this Item
Irreversible temperature-indicating liquids offer an easy, cost-effective method to verify a specific temperature on a wide range of surfaces under dynamic conditions. Paints are available in 43 different temperature ratings from 175 to 1900°F (79 to 1038°C) with accuracy within ±1% of rated temperature. They are ideal for heat treating, preventive maintenance, and any other temperature-sensitive applications. When applied to a dry smooth surface, the liquid quickly dries and forms a dull and opaque film. The film melts at the designated temperature, leaving a distinctly different mark to confirm the target temperature has been achieved. Available in 2-oz (59-mL) bottles.