Cole-Parmer Essentials Hollow Polypropylene (PP) Ball Bath Covers, 20 mm, 2000/pk
Representative image only.
Cole-Parmer Essentials Item # EW-06821-25
Reduce vapor and heat loss by 75% with one layer of balls
  • 6 Month Product / 12 Month Parts and Labor Warranty
PRODUCT OPTIONS
$363.00USD / Pkg of 2000
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionHollow Polypropylene (PP) Ball Bath Covers, 20 mm, 2000/pk
  • Warranty6 Month Product / 12 Month Parts and Labor
Key Features
  • 6 Month Product / 12 Month Parts and Labor Warranty
More About this Item
Polypropylene balls form a tight cover over liquids to prevent change in temperature and reduce evaporation. Use instead of a bath lid with any of the baths listed in the section.