Taylor 6235J 2" Dial Bi-metal Test Thermometer, 8" Stem, -10C/110C, 1% Accy

Representative image only.
Taylor Mfr # 6235J – Item # EW-90001-08
No more inaccurate temperature readings
  • Calibrate thermometer yourself for greater accuracy
  • Three-point calibration assures ±2° accuracy across entire range
  • Stainless steel 8" stem features a dimple indicating how deep to insert for the most accurate result
  • 1 year warranty

PRODUCT OPTIONS

$17.36USD / Each
$21.70USD / Each

$88.50 - $161.00USD / Each

Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Min Temperature (° F)14
  • Min Temperature (° C)-10
  • Max Temperature (° F)230
  • Max Temperature (° C)110
  • Accuracy±2° across entire range
  • Division (° C)1
  • Dial Size (in)2
  • Description2\" Dial Bi-metal Test Thermometer, 8\" Stem, -10C/110C, 1% Accy
  • Warranty1 year
1 Year Warranty

Key Features

  • Calibrate thermometer yourself for greater accuracy
  • Three-point calibration assures ±2° accuracy across entire range
  • Stainless steel 8" stem features a dimple indicating how deep to insert for the most accurate result
  • 1 year warranty

More About this Item

Models with 2" diameter dial come in a wide variety of ranges to suit more applications, and the larger size is easier to see from far away.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat